Product Name
Anti-Mouse IgG (H+L), CF™ 568 antibody produced in goat ~2 mg/mL, affinity isolated antibody
Product Number
SAB4600082
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Absorption Maximum | 562 ± 5 nm |
Degree of Labeling1 | 4-7 |
Antibody Reactivity | Mouse IgG2 |
Purity | Negligible or no free dye detected |