Product Name
Anti-Chicken IgY (IgG) (H+L), highly cross-adsorbed, CF™350 antibody produced in donkey ~2 mg/mL, affinity isolated antibody, buffered aqueous solution
Product Number
SAB4600219
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | colorless solution |
Concentration | 1.8-2.2 mg/mL |
Purity | Negligible or no free dye detected |
Reactivity | chicken IgY(IgG)(H+L) |
Absorption Maximum | 347 +/- 5 nm |
Absorption Degree of Labeling (moles dye per moles protein) | 2-6 |