Product Name
Monoclonal Anti-HLA Class I-FITC antibody produced in mouse clone W6/32, purified immunoglobulin, buffered aqueous solution
Product Number
SAB4700639
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Concentration | Report result |
Purification | The purified antibody is conjugated with Fluorescein isothiocyanate (FITC) under optimum conditions. The reagent is free of unconjugated FITC. |
Purity | N/A |
Western Blotting | Not verified |
Immunofluoresence | Not verified |
Flow Cytometry | The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1:300. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator. |