Product Name
Monoclonal Anti-MHC Class II-FITC antibody produced in rat clone M5/114, purified immunoglobulin, buffered aqueous solution
Product Number
SAB4700663
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Concentration | Report result |
Purification | The purified antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography and adjusted for direct use. No reconstitution is necessary. |
Flow cytometry | The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 2 µg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator. |