Product Name
Anti-CD11b (integrin alpha-M) antibody, Rabbit Monoclonal recombinant, expressed in HEK 293 cells, clone RM290, purified immunoglobulin
Product Number
SAB5600105
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting | 1:200-1:500 |
Immunohistochemistry | 1:100-1:200 |