Product Name
CF™ 594 succinimidyl ester suitable for fluorescence, ≥90% (TLC)
Product Number
SCJ4600031
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Reconstitution | Dissolve in DMSO |
Absorption Spectrum Λ (H2O)(nm) | 591 +/-5 nm |
Absorption Spectrum ε (408 nm) | >/= 100,000 |
Purity | >/= 90% TLC |