Product Name
Endothelin Antagonist PD 142893
Product Number
SCP0134
Product Brand
SIGMA
CAS Number
Molecular Weight
924.09
Test | Specification |
---|---|
Form | lyophilized powder |
Appearance | White to off-white |
Peptide Content | > or = to 85% |
Purity by HPLC | > or = to 95% |