Product Name
mTOR/Raptor/MLST8 human recombinant, expressed in baculovirus infected Sf9 cells, ≥50% (SDS-PAGE)
Product Number
SRP0364
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Concentration | Report result |
Specific Activity | ≥ 1.6 pmol/min/µg |
Purity (SDS-PAGE) | ≥ 50% |