Product Name
AKT1 (E17K), Active PRECISIO® Kinase, recombinant
Product Number
SRP0721
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Physical Form | Report result |
Concentration | Report result |
Purity | 70-95% |
Specific Activity | 54-74 nmol/min/mg |