Product Name
RNA Polymerase II, C-terminal human recombinant, expressed in E. coli, ≥85% (SDS-PAGE)
Product Number
SRP2013
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | frozen liquid solution |
Color | colorless and clear |
Concentration | 400 ug/mL |
Purity by SDS Page | >=85 % |