Product Name
CAR, ligand binding domain (101-348) human recombinant, expressed in E. coli, ≥85% (SDS-PAGE)
Product Number
SRP2067
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | frozen liquid solution |
Color | colorless and clear |
Concentration | 500 ug/mL |
Purity by SDS Page | >=85 % |