Product Name
IGF-Binding Protein-3 human recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Product Number
SRP3067
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report result |
Color | Report result |
Purity by SDS-Page | ≥98% |
Purity by HPLC | ≥98% |
Endotoxin Level | ≤ 1 EU/µg |
Biological Activity | Determined by its ability to inhibit IGF-II induced proliferation of MCF-7.The expected ED50 is ≤ 0.2 µg/mL in the presence of 15 ng/mL of human IGF-II |