Product Name
BMP-2 human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Product Number
SRP3326
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Appearance | Report Result |
Color | Report Result |
Purity by HPLC | ≥98% |
Purity by SDS-PAGE | ≥98% |
Endotoxin Level | < 0.1 EU/µg |
Biological Activity | Determined by its ability to induce alkaline phosphatase production by ATDC-5 cells. The expected ED50 for this effect is 0.5-1.0 µg/mL. |