Product Name
IL-6 from mouse Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Product Number
SRP3330
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Form | lyophilized powder |
Color | white to off-white |
Purity by SDS-Page | >/= 98% |
Purity by HPLC | >/= 98% |
Endotoxin Level | less than 0.1 ng per �g (1EU/�g). |
Biological Activity | Determined by its ability to stimulate the proliferation of mouse 7TD1 cells. The expected ED50 is /= 5 x 107 units/mg. |