Product Name
CHK1, active, His tagged human PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Product Number
SRP5278
Product Brand
SIGMA
CAS Number
154907-65-0
Molecular Weight
Test | Specification |
---|---|
Purity by SDS-PAGE | >= 70% |
Specific Activity | 122-190 nmol/min/mg |