Product Name
BUB1B, active, GST tagged human PRECISIO®, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Product Number
SRP5327
Product Brand
SIGMA
CAS Number
203945-19-1
Molecular Weight
Test | Specification |
---|---|
Purity by SDS-PAGE | >= 70% |
Specific Activity | 1.2-1.8 nmol/min/mg |