Product Name
PEDF mouse recombinant, expressed in CHO cells, FLAG® tagged, >95% (SDS-PAGE)
Product Number
SRP8048
Product Brand
SIGMA
CAS Number
607410-75-3
Molecular Weight
Test | Specification |
---|---|
Source | recombinant expressed in CHO cells |
Molecular Weight | ~47 kDa (SDS-PAGE) |
Storage/Stability | -20�C. Avoid repeated freeze/thaw cycles. |
Formulation | Lyophilized from PBS. |
Reconstitution | Reconstitute with 100 ul deionized water. |
Form | lyophilized powder or pellet |
Color | white |
Purity by SDS-PAGE | >=95% |
Endotoxin Level | <0.01 EU/ug purified protein (LAL test). |