Product Name
Thioglucosidase from Sinapis alba (white mustard) seed ≥100 units/g solid
Product Number
T4528
Product Brand
SIGMA
CAS Number
9025-38-1
Molecular Weight
Test | Specification |
---|---|
APPEARANCE | REPORT RESULT |
ENZYMATIC ACTIVITY | MINIMUM 100 UNITS/G SOLID |
UNIT DEFINITION | ONE UNIT WILL PRODUCE 1.0MICROMOLE OF GLUCOSE PER MINUTE FROM SINIGRIN AT PH6.0 AT 25DEGC |
DOCUMENT # | DOCUMENT # T4528/01/13/04/1 |