Product Name
Monoclonal Anti-FHIT antibody produced in mouse clone 1C3, purified immunoglobulin, buffered aqueous solution
Product Number
WH0002272M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Sandwich ELISA | Detection limit for recombinant GST tagged FHIT is approximately 0.1ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~37 kDa using immunogen protein lysate. |