Product Name
Monoclonal Anti-SHMT1 antibody produced in mouse clone 4F9, purified immunoglobulin, buffered aqueous solution
Product Number
WH0006470M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~53 kDa using HeLa lysate. |
Immunohistochemistry-Paraffin Embedded Tissue (IHC-P) | Antibody concentration: 1 ug/ml using human placenta tissue. |
Sandwich ELISA | Detection limit for recombinant GST tagged SHMT1 is approximately 1ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~38 kDa using immunogen protein lysate. |