Product Name
Monoclonal Anti-TREM1 antibody produced in mouse clone 2E2, purified immunoglobulin, buffered aqueous solution
Product Number
WH0054210M4
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 ~ 5 ug/ml. Specific band of ~26 kDa using HL-60 lysate. |
Sandwich ELISA | Detection limit for recombinant GST tagged TREM1 is approximately 0.03ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~50 kDa using immunogen protein lysate. |