Product Name
Monoclonal Anti-XPO5 antibody produced in mouse clone 2C5-1B3, purified immunoglobulin, buffered aqueous solution
Product Number
WH0057510M1
Product Brand
SIGMA
CAS Number
Molecular Weight
Test | Specification |
---|---|
Western Blotting (Cell lysate) | Antibody concentration: 1 - 5 ug/ml. Specific band of ~136 kDa using Hela NE lysate. |
Immunofluorescence (IF) | Antibody concentration: 10 ug/ml using HeLa cell. |
Sandwich ELISA | Detection limit for recombinant GST tagged XPO5 is approximately 3 ng/ml. |
ELISA (Recombinant protein) | Antibody concentration: 1 ug/ml using recombinant protein. |
Western Blotting (Recombinant protein) | Antibody concentration: 1 ug/ml. Specific band of ~57 kDa using immunogen protein lysate. |