Product Name
Dimethyl adipate analytical standard
Product Number
72849
Product Brand
SUPELCO
CAS Number
627-93-0
Molecular Weight
174.19
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 95.0 % |
REFRACTIVE INDEX N20/D | 1.426 - 1.430 |
PROTON NMR SPECTRUM | CONFORMS TO STRUCTURE |
EXPIRY PERIOD | 36 MONTHS |