Product Name
HPTLC calibration mix in methanol, analytical standard
Product Number
91816
Product Brand
SUPELCO
CAS Number
Molecular Weight
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Light Yellow |
APPEARANCE (FORM) | Liquid |
IDENTIY HPLC | CORRESPONDS TO REQUIREMENTS |
DENSITY D20/4 | 0.791 - 0.799 |
REFRACTIVE INDEX N20/D | 1.328 - 1.334 |
EXPIRY PERIOD | 36 MONTHS |