
Anti-MAP2K2/MEK2 Antibody

Synonym(s):
Anti-MAPKK, Anti-MEK, Anti-MEK2, Anti-PRKMK2
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing