
Anti-PIK3R1 antibody produced in rabbit

Synonym(s):
Anti-PI3-kinase p85-subunit α antibody produced in rabbit, Anti-PI3K antibody produced in rabbit, Anti-Phosphatidylinositol 3-kinase regulatory subunit α antibody produced in rabbit, Anti-PtdIns-3-kinase p85-α antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1