
Anti-TAF5 antibody produced in rabbit

Synonym(s):
Anti-TAF(II)100 antibody produced in rabbit, Anti-TAFII-100 antibody produced in rabbit, Anti-TAFII100 antibody produced in rabbit, Anti-Transcription initiation factor TFIID 100 kDa subunit antibody produced in rabbit, Anti-Transcription initiation factor TFIID subunit 5 antibody produced in rabbit, TAF(II)100, TAFII-100, TAFII100, Transcription initiation factor TFIID 100 kDa subunit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing