Skip to Content
MilliporeSigma
vibrant-m

Casein Acid Hydrolysate for Biotechnology and Microbiology

vibrant-m
Synonym(s):
Casein hydrolyzates
CAS Number:
EC Number:

Products

Shipping
Product Category
Biological Source
Physical Form
Tissue
Quality Segment
Application
Microorganism Suitability