
Clonazepam Related Compound C

Synonym(s):
2-Bromo-N-[2-(2-chlorobenzoyl)-4-nitrophenyl]acetamide
Empirical Formula (Hill Notation):
C15H10BrClN2O4
CAS Number:
Molecular Weight:
397.61
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing