
Difenoconazol Metabolite CGA 205375

Synonym(s):
1-[2-Chloro-4-(4-chlorophenoxy)phenyl]-2-[1,2,4]triazol-1-ylethanol
Empirical Formula (Hill Notation):
C16H13Cl2N3O2
CAS Number:
Molecular Weight:
350.20
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing