
Lipopolysaccharides from Escherichia coli O127:B8

Synonym(s):
LPS
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Color
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing