Skip to Content
US
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Manganese/nickel
Products
Manufacturer
Physical Form
Diameter (mm)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
GF57147712
Manganese/nickel,
foil, Mn 88%/Ni 12%, thickness 0.5 mm, 6 mm diameter, temper as rolled
Expand
View Pricing
GF93015851
Manganese/nickel,
foil, thickness 0.5 mm, Mn88%/Ni12%, size 25 × 25 mm, temper as rolled
Expand
View Pricing
GF22389363
Manganese/nickel,
foil, Mn 88%/Ni 12%, thickness 0.5 mm, 15 mm diameter, temper as rolled
Expand
View Pricing
GF43463831
Manganese/nickel,
foil, Mn 88%/Ni 12%, thickness 0.5 mm, 8 mm diameter, temper as rolled
Expand
View Pricing
GF82857189
Manganese/nickel,
foil, Mn88%/Ni12%, thickness 0.5 mm, diameter 4 mm, temper as rolled
Expand
View Pricing
GF32979112
Manganese/nickel,
foil, Mn 88%/Ni 12%, thickness 0.5 mm, 10 mm diameter, temper as rolled
Expand
View Pricing
GF26981309
Manganese/nickel,
foil, Mn 88%/Ni 12%, thickness 0.5 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing