Skip to Content
MilliporeSigma
Pellicon® 2 Cassette Ultrafiltration Module PLCTK V 0.5 m2 filtration area 0.5 m2, NMWCO 30 kDa

Pellicon® 2 Cassette Ultrafiltration Module PLCTK V 0.5 m2

Pellicon® 2 Cassette Ultrafiltration Module PLCTK V 0.5 m2 filtration area 0.5 m2, NMWCO 30 kDa
Synonym(s):
Pellicon® 2 Cassette Ultrafiltration Module

Products

Brand
Length (cm)
Length (in)
Quality Segment
MWCO (kDa)
Technique
Showing 1-1 of 1