
Sweet blood orange oil

Synonym(s):
Orange oil, Citrus aurantium, Citrus sinensis
CAS Number:
Products
Product Category
Brand
Sustainability
Biological Source
Feature
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing