MilliporeSigma
  • Comprehensive bioinformation analysis of the miRNA of PLCE1 knockdown in esophageal squamous cell carcinoma.
Comprehensive bioinformation analysis of the miRNA of PLCE1 knockdown in esophageal squamous cell carcinoma.
Molecular and cellular biochemistry (2017-12-15)
Xiaobin Cui, Kaige Wang, Xinqian Yang, Hao Peng, Xi Chen, Huahua Xin, Yanxia Tian, Yunzhao Chen, Feng Li
ABSTRACT
Phospholipase C epsilon 1 (PLCE1) has been recognized as a novel susceptibility marker for esophageal squamous cell carcinoma (ESCC). The purpose of our study is to investigate its effect on the regulation of miRNA expression so as to translating the data into a novel strategy in control of ESCC. In this study, PLCE1 siRNA and vector-only plasmid were stably transfected into Eca109 and EC9706 cells and then subjected to miRNA array analysis, and quantitative real-time PCR was applied to validate miRNA array data. Then bioinformatic analyses, such as GO and pathway software, were conducted to obtain data on these differentially expressed miRNAs-targeted genes (DEGs) and clarify their function and pathway. The results showed that 36 miRNAs were found to be differentially expressed in PLCE1 siRNA-transfected cells compared with the control cells. In particular, 28 miRNAs were upregulated while 8 miRNAs were downregulated. Gene Ontology analysis showed that the function of the DEGs included cell cycle arrest, cell-matrix adhesion, apoptosis, etc. After this, the major pathways associated with the DEGs were regulation of actin cytoskeleton, TGF-beta signaling pathway, Notch signaling pathway and so on. Taken together, these results showed that the knockdown of PLCE1 may play a vital role in the control of ESCC. Further investigation will reveal and verify the function and pathway of the DEGs for the development of novel treatment strategy for the better control of ESCC.
Sigma-Aldrich
MISSION® esiRNA, targeting human PLCE1