MilliporeSigma
  • All-trans-retinoic acid stimulates translation and induces spine formation in hippocampal neurons through a membrane-associated RARalpha.
All-trans-retinoic acid stimulates translation and induces spine formation in hippocampal neurons through a membrane-associated RARalpha.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2007-08-23)
Na Chen, Joseph L Napoli
ABSTRACT
Differentiation and patterning in the developing nervous system require the vitamin A metabolite all-trans-retinoic acid (atRA). Recent data suggest that higher cognitive functions, such as creation of hippocampal memory, also require atRA and its receptors, RAR, through affecting synaptic plasticity. Here we show that within 30 min atRA increased dendritic growth approximately 2-fold, and PSD-95 and synaptophysin puncta intensity approximately 3-fold, in cultured mouse hippocampal neurons, suggesting increased synapse formation. atRA (10 nM) increased ERK1/2 phosphorylation within 10 min. In synaptoneurosomes, atRA rapidly increased phosphorylation of ERK1/2, its target 4E-BP, and p70S6K, and its substrate, ribosome protein S6, indicating activation of MAPK and mammalian target of rapamycin (mTOR). Immunofluorescence revealed intense dendritic expression of RARalpha in the mouse hippocampus and localization of RARalpha on the surfaces of primary cultures of hippocampal neurons, with bright puncta along soma and neurites. Surface biotinylation confirmed the locus of RARalpha expression. Knockdown of RARalpha by shRNA impaired atRA-induced spine formation and abolished dendritic growth. Prolonged atRA stimulation reduced surface/total RARalpha by 43%, suggesting internalization, whereas brain-derived nerve growth factor or bicuculline increased the ratio by approximately 1.8-fold. atRA increased translation in the somatodendritic compartment, similar to brain-derived nerve growth factor. atRA specifically increased dendritic translation and surface expression of the alpha-amino-3-hydroxyl-5-methyl-4-isoxazole propionate receptor (AMPAR) subunit 1 (GluR1), without affecting GluR2. These data provide mechanistic insight into atRA function in the hippocampus and identify a unique membrane-associated RARalpha that mediates rapid induction of neuronal translation by atRA.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)