MilliporeSigma
Cloning of a mouse adrenocorticotropin receptor-encoding gene.
Gene (1995-02-14)
M Kubo, T Ishizuka, H Kijima, M Kakinuma, T Koike
ABSTRACT
A genomic DNA encoding a mouse adrenocorticotropin (ACTH) receptor was isolated. The predicted 296-amino-acid sequence showed 88.9 and 78.7% identities to the human and bovine homologues, respectively.