MilliporeSigma
  • Neuropilin-2 is a receptor for semaphorin IV: insight into the structural basis of receptor function and specificity.
Neuropilin-2 is a receptor for semaphorin IV: insight into the structural basis of receptor function and specificity.
Neuron (1998-12-18)
R J Giger, E R Urquhart, S K Gillespie, D V Levengood, D D Ginty, A L Kolodkin
ABSTRACT
Neuropilins bind secreted members of the semaphorin family of proteins. Neuropilin-1 is a receptor for Sema III. Here, we show that neuropilin-2 is a receptor for the secreted semaphorin Sema IV and acts selectively to mediate repulsive guidance events in discrete populations of neurons. neuropilin-2 and semaIV are expressed in strikingly complementary patterns during neurodevelopment. The extracellular complement-binding (CUB) and coagulation factor domains of neuropilin-2 confer specificity to the Sema IV repulsive response, and these domains of neuropilin-1 are necessary and sufficient for binding of the Sema III semaphorin (sema) domain. The coagulation factor domains alone are necessary and sufficient for binding of the Sema III immunoglobulin- (Ig-) basic domain and the unrelated ligand, vascular endothelial growth factor (VEGF). Lastly, neuropilin-1 can homomultimerize and form heteromultimers with neuropilin-2. These results provide insight into how interactions between neuropilins and secreted semaphorins function to coordinate repulsive axon guidance during neurodevelopment.