MilliporeSigma
  • Inhibition of the H3K4 methyltransferase MLL1/WDR5 complex attenuates renal senescence in ischemia reperfusion mice by reduction of p16INK4a.
Inhibition of the H3K4 methyltransferase MLL1/WDR5 complex attenuates renal senescence in ischemia reperfusion mice by reduction of p16INK4a.
Kidney international (2019-10-02)
Hironori Shimoda, Shigehiro Doi, Ayumu Nakashima, Kensuke Sasaki, Toshiki Doi, Takao Masaki
ABSTRACT
Renal function declines with aging and is pathologically characterized by chronic inflammation and fibrosis. Renal senescence is induced not only by aging but also by various stimuli, including ischemia reperfusion injury. Recently, the accumulation of p16INK4a-positive cells in the kidney has been considered a molecular feature of renal senescence, with the p16INK4a gene reportedly regulated by mixed-lineage leukemia 1 (MLL1)/WD-40 repeat protein 5 (WDR5)-mediated histone 3 lysine 4 trimethylation (H3K4me3). Here, we determined whether inhibition of MLL1/WDR5 activity attenuates renal senescence, inflammation, and fibrosis in mice with ischemia reperfusion injury and in cultured rat renal fibroblasts. MM-102 or OICR-9429, both MLL1/WDR5 protein-protein interaction inhibitors, and small interfering RNA (siRNA) for MLL1 or WDR5 suppressed the expression of p16INK4a in mice with ischemia reperfusion injury, accompanied by downregulation of H3K4me3 expression. MM-102 attenuated renal fibrosis and inflammation in the kidney of mice with ischemia reperfusion injury. Moreover, in vitro study showed that transforming growth factor-β1 induced the expression of MLL1, WDR5, H3K4me3, and p16INK4a. Finally, chromatin immunoprecipitation identified the p16INK4a promoter at an H3K4me3 site in renal fibroblasts. Thus, our findings show that H3K4me3 inhibition ameliorates ischemia reperfusion-induced renal senescence along with fibrosis and inflammation.
Sigma-Aldrich
Magna ChIP® A/G Chromatin Immunoprecipitation Kit, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A/G beads.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human WDR5
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid