MilliporeSigma
  • Design on a Rational Basis of High-Affinity Peptides Inhibiting the Histone Chaperone ASF1.
Design on a Rational Basis of High-Affinity Peptides Inhibiting the Histone Chaperone ASF1.
Cell chemical biology (2019-09-24)
May Bakail, Albane Gaubert, Jessica Andreani, Gwenaëlle Moal, Guillaume Pinna, Ekaterina Boyarchuk, Marie-Cécile Gaillard, Regis Courbeyrette, Carl Mann, Jean-Yves Thuret, Bérengère Guichard, Brice Murciano, Nicolas Richet, Adeline Poitou, Claire Frederic, Marie-Hélène Le Du, Morgane Agez, Caroline Roelants, Zachary A Gurard-Levin, Geneviève Almouzni, Nadia Cherradi, Raphael Guerois, Françoise Ochsenbein
ABSTRACT
Anti-silencing function 1 (ASF1) is a conserved H3-H4 histone chaperone involved in histone dynamics during replication, transcription, and DNA repair. Overexpressed in proliferating tissues including many tumors, ASF1 has emerged as a promising therapeutic target. Here, we combine structural, computational, and biochemical approaches to design peptides that inhibit the ASF1-histone interaction. Starting from the structure of the human ASF1-histone complex, we developed a rational design strategy combining epitope tethering and optimization of interface contacts to identify a potent peptide inhibitor with a dissociation constant of 3 nM. When introduced into cultured cells, the inhibitors impair cell proliferation, perturb cell-cycle progression, and reduce cell migration and invasion in a manner commensurate with their affinity for ASF1. Finally, we find that direct injection of the most potent ASF1 peptide inhibitor in mouse allografts reduces tumor growth. Our results open new avenues to use ASF1 inhibitors as promising leads for cancer therapy.
Sigma-Aldrich
Ribonuclease A from bovine pancreas, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human ASF1A
Sigma-Aldrich
Nuclease micrococcal from Staphylococcus aureus, 100-300 units/mg protein
Sigma-Aldrich
Propidium iodide solution