- Entry of the bat influenza H17N10 virus into mammalian cells is enabled by the MHC class II HLA-DR receptor.
Entry of the bat influenza H17N10 virus into mammalian cells is enabled by the MHC class II HLA-DR receptor.
Nature microbiology (2019-07-31)
Efstathios S Giotis, George Carnell, Erik F Young, Saleena Ghanny, Patricia Soteropoulos, Lin-Fa Wang, Wendy S Barclay, Michael A Skinner, Nigel Temperton
PMID31358984
ABSTRACT
Haemagglutinin and neuraminidase surface glycoproteins of the bat influenza H17N10 virus neither bind to nor cleave sialic acid receptors, indicating that this virus employs cell entry mechanisms distinct from those of classical influenza A viruses. We observed that certain human haematopoietic cancer cell lines and canine MDCK II cells are susceptible to H17-pseudotyped viruses. We identified the human HLA-DR receptor as an entry mediator for H17 pseudotypes, suggesting that H17N10 possesses zoonotic potential.