Merck
  • Divergence of Iodine and Thyroid Hormones in the Fetal and Maternal Parts of Human-Term Placenta.
Divergence of Iodine and Thyroid Hormones in the Fetal and Maternal Parts of Human-Term Placenta.
Biological trace element research (2019-09-11)
Shiqiao Peng, Chenyan Li, Xiaochen Xie, Xiaomei Zhang, Danyang Wang, Xixuan Lu, Manni Sun, Tao Meng, Shiwei Wang, Yaqiu Jiang, Zhongyan Shan, Weiping Teng
ABSTRACT
The human placenta is an important organ that forms a barrier where maternal and fetal exchange takes place. The placenta transport iodine to the fetal circulation by transfer of maternal iodine and deiodination of thyroid hormones (THs). The aim of the study was to examine the distribution of iodine and thyroid hormone transporters in the maternal and fetal sides of human-term placenta. A cross-sectional study was performed at the First Affiliated Hospital of China Medical University. Placental samples (maternal and fetal surfaces) were collected from 113 healthy-term pregnant women. The iodine content; the concentration of thyroxine (T4), triiodothyronine (T3), and reverse T3 (rT3); and the enzyme activity of placental type 2 iodothyronine deiodinase (D2) and D3 were examined. The mRNA and protein localization/expression of iodine and thyroid hormone transporters in the placenta were also studied. We also analyzed the association between expression level of Na+/I- symporter (NIS), thyroid hormone transporter protein, D3 activity in maternal and fetal surfaces of placenta with iodine content, and thyroid hormone levels. Iodine levels in placental samples from the maternal side were significantly higher than those in samples from the fetal side. T3 and T4 expression in fetal placenta was significantly lower than in maternal placenta. D3 activity in the fetal side of the placentas was significantly higher than that in the maternal side. The mRNA and protein expression of monocarboxylate transporters 8 (MCT8), L-amino acid transporters 1 (LAT1), organic anion transporting polypeptides 4A1 (OATP4A1), and TH binding protein transthyretin (TTR) were significantly increased in maternal side, while the NIS expression was higher in fetal side of human-term placenta. In conclusion, the enzymatic deiodination of thyroid hormones forms a barrier which reduces transplacental passage of the hormones and that the maternal part of the placenta is the primary factor in the mechanism regulating the hormonal transfer.