MilliporeSigma
  • Thymus‑expressed chemokine secreted by breast cancer cells promotes metastasis and inhibits apoptosis.
Thymus‑expressed chemokine secreted by breast cancer cells promotes metastasis and inhibits apoptosis.
Oncology reports (2020-04-24)
Lu Chen, Shuming Zhang, Yaqian Shen, Linzeng Qi, Zhaolin Zhang, Hua Tian, Zhigeng Zou
ABSTRACT
The aim of the present study was to investigate the underlying mechanisms of thymus‑expressed chemokine (TECK) autocrine signaling, and its effect on carcinogenesis and the development of breast cancer. The present study also assessed epithelial‑mensenchymal transition (EMT) and cell migration, invasion, proliferation and apoptosis. Breast cancer cell lines MCF‑7 and MDA‑MB‑231 were used in the present study, and TECK basic expression in cancer cells was investigated using western blotting (WB). EMT markers, Akt pathway molecules and apoptosis indicators were detected by reverse transcription‑quantitative PCR or WB. In order to assess migration and invasion, wound healing and Matrigel invasion assays were performed. Moreover, flow cytometry was used to assess the rate of proliferation and apoptosis. In vivo experiments were conducted in nude mice to assess cancer growth. It was revealed that breast cancer cells could secrete TECK in an autocrine manner. Furthermore, TECK could increase cell migration and invasion by promoting EMT and inhibit apoptosis via the Akt signaling pathway.