MilliporeSigma
  • Sequential CRISPR-Based Screens Identify LITAF and CDIP1 as the Bacillus cereus Hemolysin BL Toxin Host Receptors.
Sequential CRISPR-Based Screens Identify LITAF and CDIP1 as the Bacillus cereus Hemolysin BL Toxin Host Receptors.
Cell host & microbe (2020-06-17)
Jie Liu, Zehua Zuo, Inka Sastalla, Chengyu Liu, Ji Yong Jang, Yusuke Sekine, Yuesheng Li, Mehdi Pirooznia, Stephen H Leppla, Toren Finkel, Shihui Liu
ABSTRACT
Bacteria and their toxins are associated with significant human morbidity and mortality. While a few bacterial toxins are well characterized, the mechanism of action for most toxins has not been elucidated, thereby limiting therapeutic advances. One such example is the highly potent pore-forming toxin, hemolysin BL (HBL), produced by the gram-positive pathogen Bacillus cereus. However, how HBL exerts its effects and whether it requires any host factors is unknown. Here, we describe an unbiased genome-wide CRISPR-Cas9 knockout screen that identified LPS-induced TNF-α factor (LITAF) as the HBL receptor. Using LITAF-deficient cells, a second, subsequent whole-genome CRISPR-Cas9 screen identified the LITAF-like protein CDIP1 as a second, alternative receptor. We generated LITAF-deficient mice, which exhibit marked resistance to lethal HBL challenges. This work outlines and validates an approach to use iterative genome-wide CRISPR-Cas9 screens to identify the complement of host factors exploited by bacterial toxins to exert their myriad biological effects.
Sigma-Aldrich
Ethyl methanesulfonate, liquid
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Gelatin solution, Type B, 2% in H2O, tissue culture grade, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)