- Soluble PD-L1 improved direct ARDS by reducing monocyte-derived macrophages.
Soluble PD-L1 improved direct ARDS by reducing monocyte-derived macrophages.
Acute respiratory distress syndrome (ARDS) is common in intensive care units (ICUs), although it is associated with high mortality, no effective pharmacological treatments are currently available. Despite being poorly understood, the role of programmed cell death protein 1 (PD-1) and PD-ligand 1 (PD-L1) axis in ARDS may provide significant insights into the immunosuppressive mechanisms that occur after ARDS. In the present study, we observed that the level of soluble PD-L1 (sPD-L1), a potential activator of the PD-1 pathway, was upregulated in survivors of direct ARDS than in non-survivors. Administration of sPD-L1 in mice with direct ARDS relieved inflammatory lung injury and improved the survival rate, indicating the protective role of sPD-L1 in direct ARDS. Using high-throughput mass cytometry, we found a marked decrease in the number of lung monocyte-derived macrophages (MDMs) with proinflammatory markers, and the protective role of sPD-L1 diminished in ARDS mice with monocyte/macrophage depletion. Furthermore, PD-1 expression increased in the MDMs of patients and mice with direct ARDS. Finally, we showed that sPD-L1 induced MDM apoptosis in patients with direct ARDS. Taken together, our results demonstrated that the engagement of sPD-L1 on PD-1 expressing macrophages resulted in a decrease in pro-inflammatory macrophages and eventually improved direct ARDS. Our study identified a prognostic indicator for patients with direct ARDS and a potential target for therapeutic development in direct ARDS.