MilliporeSigma
  • Enhanced psychostimulant response, but not social avoidance, depends on GluA1 AMPA receptors in VTA dopamine neurons following intermittent social defeat stress in rats.
Enhanced psychostimulant response, but not social avoidance, depends on GluA1 AMPA receptors in VTA dopamine neurons following intermittent social defeat stress in rats.
The European journal of neuroscience (2020-07-01)
Megan L Rudolph, Racheal L Neve, Ronald P Hammer, Ella M Nikulina
ABSTRACT
Evidence from both human and animal studies demonstrates the importance of social stress in the development of addiction-related behaviour. In rats, intermittent social defeat stress causes long-lasting psychostimulant cross-sensitization. Our recent data reveal heightened expression of AMPA receptor (AMPAR) GluA1 subunit in rat ventral tegmental area (VTA), which occurs concurrently with social stress-induced amphetamine (AMPH) cross-sensitization. In addition, social stress in rats induced social avoidance behaviour. The present study evaluated the effects of intermittent social defeat stress on GluA1 expression in VTA dopamine (DA) neurons, then utilized Cre-dependent virus-mediated gene transfer to determine the functional role of homomeric GluA1-AMPARs in these neurons. Social defeat stress exposure induced GluA1 expression in VTA DA neurons, as demonstrated by a greater density of GluA1/tyrosine hydroxylase (TH) double-labelling in VTA neurons in stressed rats. Additionally, functional inactivation of VTA GluA1 AMPARs in DA neurons prevented stress-induced cross-sensitization, or augmented locomotor response to low dose AMPH challenge (1.0 mg/kg, i.p.), but had no effect on social stress-induced social avoidance behaviour. Furthermore, wild-type overexpression of GluA1 in VTA DA neurons had the opposite effect; locomotor-activating effects of AMPH were significantly augmented, even in the absence of stress. Taken together, these results suggest that stress-induced GluA1 expression in VTA DA neurons is necessary for psychostimulant cross-sensitization, but not for social avoidance. This differential effect suggests that different neural pathways are implicated in these behaviours. These findings could lead to novel pharmacotherapies to help prevent stress-induced susceptibility to substance abuse.
Sigma-Aldrich
Anti-GluR1 Antibody, from rabbit, purified by affinity chromatography