MilliporeSigma
Cecr2 mutant mice as a model for human cat eye syndrome.
Scientific reports (2021-02-06)
Renée Dicipulo, Kacie A Norton, Nicholas A Fairbridge, Yana Kibalnyk, Sabrina C Fox, Lisa K Hornberger, Heather E McDermid
ABSTRACT
Cat eye syndrome (CES), a human genetic disorder caused by the inverted duplication of a region on chromosome 22, has been known since the late 1890s. Despite the significant impact this disorder has on affected individuals, models for CES have not been produced due to the difficulty of effectively duplicating the corresponding chromosome region in an animal model. However, the study of phenotypes associated with individual genes in this region such as CECR2 may shed light on the etiology of CES. In this study we have shown that deleterious loss of function mutations in mouse Cecr2 effectively demonstrate many of the abnormal features present in human patients with CES, including coloboma and specific skeletal, kidney and heart defects. Beyond phenotypic analyses we have demonstrated the importance of utilizing multiple genetic backgrounds to study disease models, as we see major differences in penetrance of Cecr2-related abnormal phenotype between mouse strains, reminiscent of the variability in the human syndrome. These findings suggest that Cecr2 is involved in the abnormal features of CES and that Cecr2 mice can be used as a model system to study the wide range of phenotypes present in CES.