MilliporeSigma
Autophagy mediates the mitotic senescence transition.
Genes & development (2009-03-13)
Andrew R J Young, Masako Narita, Manuela Ferreira, Kristina Kirschner, Mahito Sadaie, Jeremy F J Darot, Simon Tavaré, Satoko Arakawa, Shigeomi Shimizu, Fiona M Watt, Masashi Narita
ABSTRACT
As a stress response, senescence is a dynamic process involving multiple effector mechanisms whose combination determines the phenotypic quality. Here we identify autophagy as a new effector mechanism of senescence. Autophagy is activated during senescence and its activation is correlated with negative feedback in the PI3K-mammalian target of rapamycin (mTOR) pathway. A subset of autophagy-related genes are up-regulated during senescence: Overexpression of one of those genes, ULK3, induces autophagy and senescence. Furthermore, inhibition of autophagy delays the senescence phenotype, including senescence-associated secretion. Our data suggest that autophagy, and its consequent protein turnover, mediate the acquisition of the senescence phenotype.