MilliporeSigma
Transiently transfected purine biosynthetic enzymes form stress bodies.
PloS one (2013-02-14)
Alice Zhao, Mark Tsechansky, Jagannath Swaminathan, Lindsey Cook, Andrew D Ellington, Edward M Marcotte
ABSTRACT
It has been hypothesized that components of enzymatic pathways might organize into intracellular assemblies to improve their catalytic efficiency or lead to coordinate regulation. Accordingly, de novo purine biosynthesis enzymes may form a purinosome in the absence of purines, and a punctate intracellular body has been identified as the purinosome. We investigated the mechanism by which human de novo purine biosynthetic enzymes might be organized into purinosomes, especially under differing cellular conditions. Irregardless of the activity of bodies formed by endogenous enzymes, we demonstrate that intracellular bodies formed by transiently transfected, fluorescently tagged human purine biosynthesis proteins are best explained as protein aggregation.