MilliporeSigma
  • Insulin-sensitizing therapy attenuates type 2 diabetes-mediated mammary tumor progression.
Insulin-sensitizing therapy attenuates type 2 diabetes-mediated mammary tumor progression.
Diabetes (2009-12-05)
Yvonne Fierz, Ruslan Novosyadlyy, Archana Vijayakumar, Shoshana Yakar, Derek LeRoith
ABSTRACT
Type 2 diabetes increases breast cancer risk and mortality, and hyperinsulinemia has been identified as a major factor linking these two diseases. Thus, we hypothesized that pharmacological reduction of elevated insulin levels would attenuate type 2 diabetes-mediated mammary tumor progression. We studied mammary tumor development in MKR(+/+) mice, a nonobese, hyperinsulinemic mouse model of type 2 diabetes. MKR(+/+) mice were either crossed with mice expressing the polyoma virus middle T oncogene specifically in the mammary gland or inoculated orthotopically with the mouse mammary tumor cell lines Met-1 and MCNeuA. MKR(+/+) or control mice harboring tumors were treated with CL-316243, a specific beta3-adrenergic receptor agonist, which sensitizes insulin action but has no direct effect on the mouse mammary epithelium or Met-1 and MCNeuA cells. CL-316243 treatment significantly reduced the elevated insulin levels in MKR(+/+) mice and, as a consequence, attenuated mammary tumor progression in the three tumor models tested. This effect was accompanied by reductions in phosphorylation of insulin and IGF-I receptors in transformed mammary tissue. Insulin-sensitizing treatment is sufficient to abrogate type 2 diabetes-mediated mammary tumor progression. Therefore, early administration of insulin-sensitizing therapy may reduce breast cancer risk and mortality in patients with type 2 diabetes.
Sigma-Aldrich
CL 316,243 hydrate, ≥98% (HPLC), powder